Learn More
Sino Biological Anti-ADAM15 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10517MM11P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ADAM15 (rh ADAM15; Catalog#10517-H08H; NP_997074.1; Met1-Thr696) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8846