Learn More
Sino Biological Recombinant Anti-ADAM15 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10517R007A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ADAM15 (rh ADAM15; Catalog#10517-H08H; NP_997074.1; Met 1-Thr 696) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-8183