Learn More
Sino Biological Anti-ADAM15 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10517RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ADAM15 (rh ADAM15; Catalog#10517-H08H; NP_997074.1; Met 1-Thr 696). ADAM15 specific IgG was purified by human ADAM15 affinity chromatography.
Catalog No. 50-240-8380