Learn More
Sino Biological Anti-ADAM17 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 200058T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human ADAM17 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3979