Learn More
ACROBiosystems ACROBiosystems Human ADAM8 / CD156a Protein, His Tag
Catalog No. 502106272
Encompass_Preferred
Recombinant Protein;500UG;Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession # AAI15405.1).;Human ADAM8, His Tag fused with a polyhistidine tag at the C-terminus, has a calculated MW of 53.8 kDa, which migrates as a 130-150 kDa band in DTT-reduced SDS-PAGE due to glycosylation. Two main fragments bands are visible with apparent MW of 58-68 kDa and 35-40 kDa respectively in reduced SDS-PAGE as a result of its being prone to proteolytic cleavage.
Catalog No. 50-210-6272 Supplier ACROBiosystems Supplier No. AD8H5223500UG
Recombinant Protein;500UG;Human ADAM8, His Tag (AD8-H5223) is expressed from human 293 cells (HEK293). It contains AA Ile 17 - Pro 497 (Accession # AAI15405.1).;Human ADAM8, His Tag fused with a polyhistidine tag at the C-terminus, has a calculated MW of 53.8 kDa, which migrates as a 130-150 kDa band in DTT-reduced SDS-PAGE due to glycosylation. Two main fragments bands are visible with apparent MW of 58-68 kDa and 35-40 kDa respectively in reduced SDS-PAGE as a result of its being prone to proteolytic cleavage.