Learn More
Raybiotech Inc Human ADAM-9 ELISA Kit
Supplier:  Raybiotech Inc ELHADAM91
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human ADAM9. Other species not determined yet.
Catalog No. 50-194-8950