Learn More
ACROBiosystems Human ADAM9, His Tag (AD9-H52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 206 - Asp 697 (Accession # Q13443-1).
Supplier:  ACROBiosystems AD9H52H750UG
Encompass_Preferred
ADAM9 (A disintegrin and a metalloprotease 9) is a membrane-anchored protein that participates in a variety of physiological functions, primarily through the disintegrin domain for adhesion and the metalloprotease domain for ectodomain shedding of a wide variety of cell surface proteins. ADAM9 influences the developmental process, inflammation, and degenerative diseases.Recently, increasing evidence has shown that ADAM9 plays an important role in tumor biology. Overexpression of ADAM9 has been found in several cancer types and is correlated with tumoraggressiveness and poor prognosis. In addition, through either proteolytic or non-proteolytic pathways, ADAM9 promotes tumor progression, therapeutic resistance, and metastasis of cancers.Therefore, comprehensively understanding the mechanism of ADAM9 is crucial for the development of therapeutic anti-cancer strategies.
Catalog No. 50-223-0851