Learn More
Raybiotech Inc Human ADAMTS-13 ELISA Kit
Supplier:  Raybiotech Inc ELHADAMTS131
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 7.5 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human ADAMTS-13.
Catalog No. 50-194-8951