Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAMTS-15 ELISA Kit

Catalog No. EH13RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH13RBX5 5 x 96 Tests
EH13RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH13RBX5 Supplier Invitrogen™ Supplier No. EH13RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human ADAMTS-15 quantitates human ADAMTS-15 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human ADAMTS-15.

ADAMTS proteases are secreted enzymes containing a prometalloprotease domain of the reprolysin type. The ADAMTS proteases function in processing of procollagens and von Willebrand factor as well as catabolism of aggrecan, versican and brevican. They have been demonstrated to have important roles in connective tissue organization, coagulation, inflammation, arthritis, angiogenesis and cell migration. The immunizing peptide sequence is 100% conserved between human, mouse and rat.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TE58
Assay Range 0.45-100 ng/mL
Assay Sensitivity 0.45 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAM metallopeptidase with thrombospondin type 1 motif, 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domainsa disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 15; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 15, preproprotein; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 15; ADAM metallopeptidase with thrombospondin type 1 motif, 15; ADAMs; ADAM-TS 15; Adamts15; ADAM-TS15; ADAMTS-15; metalloendopeptidases; metalloprotease disintegrin 15 with thrombospondin domains;
Gene ID (Entrez) 170689
Gene Symbol ADAMTS15
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target ADAMTS15
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.