Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADAMTS19 (aa 664-751) Control Fragment Recombinant Protein

Catalog No. RP106856
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106856 Supplier Invitrogen™ Supplier No. RP106856
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66170. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has high sequence similarity to the protein encoded by ADAMTS16, another family member.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8TE59
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 171019
Name Human ADAMTS19 (aa 664-751) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4831407I23Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 19; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 19; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 19; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 19; ADAM metallopeptidase with thrombospondin type 1 motif, 19; ADAMs; ADAM-TS 19; Adamts19; ADAM-TS19; ADAMTS-19; AU015154; D230034E10Rik; metalloendopeptidases
Common Name ADAMTS19
Gene Symbol ADAMTS19
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DSEARDCNGPRKQYRICENPPCPAGLPGFRDWQCQAYSVRTSSPKHILQWQAVLDEEKPCALFCSPVGKEQPILLSEKVMDGTSCGYQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.