Learn More
Raybiotech Inc Human ADAMTS-4 ELISA Kit
Supplier:  Raybiotech Inc ELHADAMTS41
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human ADAMTS-4
Catalog No. 50-194-8953