Learn More
Biomatik Corporation Human Adaptor Related Protein Complex 3 Beta 1 (AP3b1) ELISA Kit
Supplier:  Biomatik Corporation EKU09227
Encompass_Preferred
Human Adaptor Related Protein Complex 3 Beta 1 (AP3b1) ELISA Kit for detecting AP3b1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00203. Gene ID: 8546. Target Synonyms: HPS; PE; HPS2; ADTB3; ADTB3A; Clathrin assembly protein complex 3 beta-1 large chain; Adaptor protein complex AP-3 subunit beta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8549