Learn More
Biomatik Corporation Human Adaptor Related Protein Complex 2 Mu 1 (AP2m1) ELISA Kit
Supplier:  Biomatik Corporation EKU02124
Encompass_Preferred
Human Adaptor Related Protein Complex 2 Mu 1 (AP2m1) ELISA Kit for detecting AP2m1 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96CW1. Gene ID: 1173. Target Synonyms: AP50; CLAPM1; Clathrin-Associated/Assembly/Adaptor Protein,Medium 1; Plasma Membrane Adaptor AP-2 50kDA Protein; Clathrin Coat Adaptor Protein AP50. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5704