Learn More
Sino Biological Human ADCYAP1R1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 201908T02100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PACAP receptor/ADCYAP1R1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-1327