Learn More
Biomatik Corporation Human Adenomatosis Polyposis Coli Protein (APC) ELISA Kit
Supplier:  Biomatik Corporation EKU02126
Encompass_Preferred
Human Adenomatosis Polyposis Coli Protein (APC) ELISA Kit for detecting APC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25054. Gene ID: 324. Target Synonyms: DP2; DP2.5; DP3; FAP; FPC; GS; Deleted in polyposis 2.5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4440