Learn More
Biomatik Corporation Human Adenosine Monophosphate Deaminase 2 (AMPD2) ELISA Kit
Supplier:  Biomatik Corporation EKU10608
Encompass_Preferred
Human Adenosine Monophosphate Deaminase 2 (AMPD2) ELISA Kit for detecting AMPD2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8,000pg/ml. Sensitivity: 55pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01433. Gene ID: 271. Target Synonyms: AMPD Isoform L. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8327