Learn More
ATCC Human adenovirus 1; Strain: Adenoid 71
Supplier:  ATCC VR1
Encompass
Human adenovirus 1 strain Adenoid 71 is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the adenoid tissue of a 5-year-old child in Maryland in 1953 and was deposited by the National Institute of Allergy and Infectious Diseases. This strain can be used in respiratory disease research.
Catalog No. 50-238-3305