Learn More
ATCC Human adenovirus 11; Strain: Slobitski
Supplier:  ATCC VR12
Encompass
Human adenovirus 11 strain Slobitski was isolated in Massachusetts from human feces. This animal virus is propagated in KB cells (ATCC CCL-17) and has applications in respiratory research.
Catalog No. 50-238-3670