Learn More
ATCC Human adenovirus 26; Strain: BP-2
Supplier:  ATCC VR3374
Encompass
Human adenovirus 26 strain BP-2 is propagated in A549 cells (ATCC CCL-185). This strain was derived from ATCC VR-1104, which was originally isolated from an anal specimen from a 9-month-old male child in Washington, DC. This product has applications in respiratory and infectious disease research.
Catalog No. 50-238-5049