Learn More
ATCC Human adenovirus 3; Strain: GB; DNA
Supplier:  ATCC VR847D
Encompass
Genomic DNA isolated from A549 cells (ATCC CCL-185) infected with Human adenovirus 3 strain G.B. (ATCC VR-847). This product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-1884