Learn More
ATCC Human adenovirus 41; Strain: Tak (73-3544)
Supplier:  ATCC VR930
Encompass
Human adenovirus 41 strain Tak (73-3544) is propagated in 293 [HEK-293] cells (ATCC CRL-1573). This strain was isolated in 1973 from the feces of a 14-month old human with gastroenteritis and was deposited by the National Institute for Public Health and the Environment, Netherlands. This product is whole-genome sequenced has applications in enteric and infectious disease research.
Catalog No. 50-238-2484