Learn More
ATCC Human adenovirus 5 strain Adenoid 75 (ATCC VR-5™)
Supplier:  ATCC VR5
Encompass
Human adenovirus 5 strain Adenoid 75 (ATCC VR-5™) is a whole-genome sequenced virus that was isolated in 1953 from the adenoid tissue of a 4-year-old girl with enlarged, chronically infected tonsils. This strain has applications in infectious disease research, respiratory studies, and assay development. It is also suitable for use in cell-based assays and molecular virology applications.
Recommended companion products:
Genomic DNA (Fisher Cat. No. 50-238-5106, ATCC VR-5DQ™)
HeLa cells (Fisher Cat. No. 50-238-3230, ATCC CCL-2™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™)
FBS (Fisher Cat. No. 50-238-3771, ATCC 30-2020™).
Catalog No. 50-238-2424