Learn More
ATCC Human adenovirus 5; Strain: Adenoid 75; DNA
Catalog No. 502384881
Encompass
Genomic DNA isolated from A549 cells (ATCC CCL-185) infected with Human adenovirus 5 (HAdV-5) strain Adenoid 75 (ATCC VR-5). This product is suitable for use in polymerase chain reaction (PCR) for viral gene products and other molecular virology applications.
Catalog No. 50-238-4881 Supplier ATCC Supplier No. VR5D
Genomic DNA isolated from A549 cells (ATCC CCL-185) infected with Human adenovirus 5 (HAdV-5) strain Adenoid 75 (ATCC VR-5). This product is suitable for use in polymerase chain reaction (PCR) for viral gene products and other molecular virology applications.