Learn More
ATCC Human adenovirus 6; Strain: Tonsil 99
Catalog No. 502384871
Encompass
Human adenovirus 6 strain Tonsil 99 is propagated in HeLa cells (ATCC CCL-2). This strain was isolated from the tonsil tissue of a 7-year-old girl with chronically infected and enlarged tonsils in Washington, DC. The strain has applications in respiratory disease research.
Catalog No. 50-238-4871 Supplier ATCC Supplier No. VR6
Human adenovirus 6 strain Tonsil 99 is propagated in HeLa cells (ATCC CCL-2). This strain was isolated from the tonsil tissue of a 7-year-old girl with chronically infected and enlarged tonsils in Washington, DC. The strain has applications in respiratory disease research.