Learn More
ATCC Human adenovirus 7; Strain: Gomen; DNA
Catalog No. 502382159
Encompass
Genomic DNA isolated from A549 cells (ATCC CCL-185) infected with Human adenovirus 7 strain Gomen (ATCC VR-7). This product has applications in PCR and other molecular biology applications.
Catalog No. 50-238-2159 Supplier ATCC Supplier No. VR7D
Genomic DNA isolated from A549 cells (ATCC CCL-185) infected with Human adenovirus 7 strain Gomen (ATCC VR-7). This product has applications in PCR and other molecular biology applications.