Learn More
Sino Biological Anti-CD97 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11280MM03P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD97 (rh CD97; Catalog#11280-H08H; NP_001775.2; Met 1-Gln 398) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4736