Learn More
Sino Biological Recombinant Anti-CD97 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 11280R024A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD97 (rh CD97; Catalog#11280-H08H; NP_001775.2; Met 1-Gln 398) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-4729