Learn More
Biomatik Corporation Human ADH (Antidiuretic Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF58486
Encompass_Preferred
Human ADH (Antidiuretic Hormone) ELISA Kit for detecting ADH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20pg/ml. Sensitivity: 0.188pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: P01185. Target Synonyms: ADH; VP; AVP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5165