Learn More
Biomatik Corporation Human Adiponectin, Adp Elisa Kit
Supplier:  Biomatik Corporation EKC31131
Encompass_Preferred
Human Adiponectin, Adp Elisa Kit for detecting Adiponectin, C1Q And Collagen Domain Containing in serum, plasma, cell culture supernates, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 1.562-100ng/ml. Sensitivity: 1.102ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q15848; ADIPOQ. Target Synonyms: 30 kDa adipocyte complement related protein; 30 kDa adipocyte complement-related protein; ACDC; Acrp 30; ACRP30; ADIPO_HUMAN; Adipocyte; Adipocyte C1q and collagen domain containing protein; Adipocyte complement related 30 kDa protein; Adipocyte complement related protein of 30 kDa; Adipocyte complement-related 30 kDa protein; adipocyte-specific secretory protein; Adiponectin; Adiponectin precursor; adiponectin; C1Q and collagen domain containing; Adipoq. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5456