Learn More
Sino Biological Recombinant Human Adiponectin Protein (hFc Tag)
Supplier:  Sino Biological 10772H02H1
Encompass_Preferred
A DNA sequence encoding the human ADIPOQ (Q15848) (Met1-Asn244) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5174