Learn More
Sino Biological Human ADK PAb 100ul (Sino Biological)
Supplier:  Sino Biological 13149RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ADK (rh ADK; Catalog#13149-H20B; AAH03568.1; Met 1-His 345). ADK specific IgG was purified by Human ADK affinity chromatography.
Catalog No. 50-237-3950