Learn More
American Research Products Inc Human ADP ribosylation factor
Catalog No. 5000327316
Encompass
Human ADP-ribosylation factor-like protein 8B (ARL8B) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27316 Supplier American Research Products Inc Supplier No. CSBEL002100HU
Human ADP-ribosylation factor-like protein 8B (ARL8B) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich