Learn More
Biomatik Corporation Human ADP Ribosylation Factor Like Protein 15 (ARL15) ELISA Kit
Supplier:  Biomatik Corporation EKU08439
Encompass_Preferred
Human ADP Ribosylation Factor Like Protein 15 (ARL15) ELISA Kit for detecting ARL15 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NXU5. Gene ID: 54622. Target Synonyms: ARFRP2; ADP-Ribosylation Factor Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5507