Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ADPRM (aa 175-246) Control Fragment Recombinant Protein

Catalog No. RP93419
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP93419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP93419 Supplier Invitrogen™ Supplier No. RP93419
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54740 (PA5-54740. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ADPRM (ADP-ribose/CDP-alcohol diphosphatase) is a protein that belongs to the Nudix hydrolase family. It plays a crucial role in the metabolism of nucleotide sugars and nucleotide sugar alcohols, which are important for glycosylation reactions in cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q3LIE5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56985
Name Human ADPRM (aa 175-246) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310004I24Rik; ADPRibase-Mn; ADP-ribose/CDP-alcohol diphosphatase, manganese dependent; ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent; ADP-ribose/CDP-alcohol pyrophosphatase; Adprm; C17orf48; CDP-choline phosphohydrolase; liver manganese (II)-dependent ADP-ribose/CDP-alcohol pyrophosphatase; manganese-dependent ADP-ribose/CDP-alcohol diphosphatase; Manganese-dependent ADP-ribose/CDP-alcohol diphosphatase-like protein; MDS006; NBLA03831; RGD1309906
Common Name ADPRM
Gene Symbol ADPRM
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QSSPKYEQCMKILREHNPNTELNSPQGLSEPQFVQFNGGFSQEQLNWLNEVLTFSDTNQEKVVIVSHLPIYP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.