Learn More
Biomatik Corporation Human Adrenergic Receptor Beta Kinase 2 (ADRbK2) ELISA Kit
Supplier:  Biomatik Corporation EKU12500
Encompass_Preferred
Human Adrenergic Receptor Beta Kinase 2 (ADRbK2) ELISA Kit for detecting ADRbK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35626. Gene ID: 157. Target Synonyms: B-ARK2; bARK2; Beta-ARK2; GRK3; G-protein-coupled receptor kinase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6093