Learn More
Biomatik Corporation Human Adrenomedullin (ADM) ELISA Kit
Supplier:  Biomatik Corporation EKU11666
Encompass_Preferred
Human Adrenomedullin (ADM) ELISA Kit for detecting ADM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35318. Gene ID: 133. Target Synonyms: AM; PAMP; Proadrenomedullin N-20 terminal peptide; ProAM N-terminal 20 peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8101