Learn More
Sino Biological Anti-AFM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13231RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AFM / Afamin (rh AFM / Afamin ; Catalog#13231-H08H; P43652; Met1-Asn599). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4269