Learn More
Proteintech Group Inc Human AFP ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE0013296T
Encompass_Preferred
AFP (alpha-fetoprotein) is a major plasma protein found in the fetus while plasma levels decrease rapidly after birth. AFP is one of the earliest markers of the hepatocyte lineage. High AFP concentrations have been correlated with tumor cell growth. Detection of AFP in plasma is important in diagnosis of hepatocellular carcinoma (HCC), stomach cancer and germ cell cancers. Altered levels of both fetal and maternal AFP have been associated with hypothyroidism, autoimmune disorders, and heart defects.
Catalog No. 50-197-4091