Learn More
Sino Biological Human AFP ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502372105
Encompass_Preferred
Each vial contains 100 ng of recombinant Human AFP / alpha-fetoprotein. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 1000 pg/mL is recommended.
Catalog No. 50-237-2105 Supplier Sino Biological Supplier No. SEK1217715
Each vial contains 100 ng of recombinant Human AFP / alpha-fetoprotein. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 1000 pg/mL is recommended.