Learn More
Sino Biological Human AFP MAb 20ul (Sino Biological)
Supplier:  Sino Biological 12177R00320
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human AFP / alpha-fetoprotein (rh AFP / alpha-fetoprotein; Catalog#12177-H08H; P02771; Met1-Val609).
Catalog No. 50-237-3819