Learn More
Sino Biological Anti-Alpha-fetoprotein Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12177MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human AFP / alpha-fetoprotein (rh AFP / alpha-fetoprotein; Catalog#12177-H08H; P02771; Met 1-Val 609). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-4434