Learn More
Sino Biological Anti-Alpha-fetoprotein Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12177RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human AFP / alpha-fetoprotein (rh AFP; Catalog#12177-H08H; P02771; Met1-Val609). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4424