Learn More
Sino Biological Anti-RAGE Antibody, Mouse Monoclonal
Supplier:  Sino Biological 11629MM02100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human RAGE/AGER (rh RAGE/AGER; Catalog#11629-H08H; NP_001127; Met1-Ala344). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-2045