Learn More
Sino Biological Recombinant Human RAGE Protein (hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11629H41HB20
Encompass_Preferred
A DNA sequence encoding the Human AGER (NP_001127)(Met1-Ala344) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4659