Learn More
Sino Biological Recombinant Human RAGE Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11629H49HB20
Encompass_Preferred
A DNA sequence encoding the Human AGER (NP_001127) (Met1-Ala344) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4657