Learn More
Biomatik Corporation Human Aggrecan (AGC) CLIA Kit
Supplier:  Biomatik Corporation EKU09927
Encompass_Preferred
Human Aggrecan (AGC) CLIA Kit for detecting AGC in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 16.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P16112. Gene ID: 176. Target Synonyms: ACAN; AGC1; AGCAN; CSPG1; CSPCP; CSPGCP; MSK16; SEDK; Cartilage-specific proteoglycan core protein; Large Aggregating Proteoglycan; Chondroitin Sulfate Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6684