Learn More
Raybiotech Inc Human Aggrecan ELISA Kit
Supplier:  Raybiotech Inc ELHACAN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 41 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Aggrecan.
Catalog No. 50-194-8940