Learn More
Sino Biological Human AGRP Protein 50ug (Sino Biological)
Supplier:  Sino Biological 10070H02H50
Encompass_Preferred
A DNA sequence encoding the human AgRP (NP_001129.1) (Met 1-Thr 132) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2550