Learn More
Sino Biological Human AGT MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10994MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiotensinogen / SerpinA8 (rh Angiotensinogen / SerpinA8; Catalog#10994-H08H; NP_000020.1; Met1-Ala485). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1201