Learn More
Sino Biological Anti-Angiotensinogen Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10994RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SerpinA8 / angiotensinogen (rh SerpinA8; Catalog#10994-H08H; NP_000020.1; Met 1-Ala 485). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2352